Learn More
Thermo Scientific™ Oxoid™ V Factor Discs
For identification of Haemophilus sp.
Brand: Thermo Scientific™ DD0004B
90.26 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Order Info
50 Discs/Cartridge
Specifications
Haemophilus species | |
V Factor Disc |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6191447 | Certificate of Analysis | 19/12/2024 | DD0004B |
6196812 | Certificate of Analysis | 19/12/2024 | DD0004B |
6183628 | Certificate of Analysis | 13/12/2024 | DD0004B |
6116048 | Certificate of Analysis | 14/06/2024 | DD0004B |
3800217 | Certificate of Analysis | 09/05/2024 | DD0004B |
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ V Factor Discs > 5 x 50 discs