Learn More
Brady™ Thermal Transfer Printer Labels: Rectangular
Rectangular printable labels for thermal transfer printers.
Brand:  Brady™ 121009
 View more versions of this product
Product Code. 17130161
  • € 236.00 / Pack of 2000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Printer Compatibility
  • BBP81
  • Brady 1244
  • Brady 1344
  • Brady 200MVP Plus
  • Brady 2461
  • Brady 300MVP Plus
  • Brady 300X-Plus II
  • Brady 3481
  • Brady 360X-Plus II
  • Brady 600X-Plus II
  • Brady 6441
  • Brady IP
  • Tagus T200
  • Tagus T300
  • Thermal Transfer Printers
This material is UL recognized with its respective ribbon; use Brady Ribbon Part# R4902.
Order Info
See the Recommended Ribbon Series for the appropriate ribbon to use.
Specifications
Specifications
Thermal Transfer Printer Labels - 3 in. Core
12.70 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Thermal Transfer Printer Labels: Rectangular > Size: 12.7W x 50.8mm H; 3 in. Core