missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL Tapered Stopper
Tapered Stoppers have excellent chemical and temperature resistance, making them an ideal laboratory accessory.
Brand: KARTELL 0145800
This item is not returnable.
View return policy
Description
- Excellent chemical and temperature resistance.
- Self-lubricating.
- Standard taper: 1:10.
- According to DIN Standard 12242.
- SUITABLE FOR FOODSTUFF.
- MATERIAL: PTFE
Specifications
Autoclavable | |
25.8 mm | |
PTFE | |
Non-sterile |
White | |
29 mm | |
Tapered |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction