Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ PTFE Support Screen Gasket
The Kimble™ Ultra-Ware™ PTFE Support Screen Gasket is suitable for use with Kimble™ Ultra-Ware™ Microfiltration Assemblies.
Brand:  DWK Life Sciences 953712-0000
Product Code. 11888532
  • € 77.50 / Pack of 25
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Product Type: Support Screen Gasket
  • Material: PTFE
  • Size: 25 mm
  • For Use with: Kimble™ Ultra-Ware™ Microfiltration Assembly
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
Kimble Ultra-Ware Microfiltration Assembly
25 Pk.
25 mm
Reusable
PTFE
Gasket
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers