missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX3 Mouse anti-Human, Rat, Clone: 1B9C7, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | STX3 |
|---|---|
| Clone | 1B9C7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828935
|
Proteintech
66760-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818935
|
Proteintech
66760-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The gene is a member of the syntaxin family. The encoded protein is targeted to the apical membrane of epithelial cells where it forms clusters and is important in establishing and maintaining polarity necessary for protein trafficking involving vesicle fusion and exocytosis. Alternative splicing results in multiple transcript variants.Specifications
| STX3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| STX3, STX3A, syntaxin 3 | |
| Stx3 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B9C7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat, Human | |
| Q08849, Q13277 | |
| 6809, 81802 | |
| STX3 Fusion Protein Ag7805 | |
| Primary | |
| -20°C | |
| STX3 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto