missing translation for 'onlineSavingsMsg'
Learn More

STX3 Mouse anti-Human, Rat, Clone: 1B9C7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66760-1-IG-150UL

 View more versions of this product

Product Code. 16818935

  • € 410.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The gene is a member of the syntaxin family. The encoded protein is targeted to the apical membrane of epithelial cells where it forms clusters and is important in establishing and maintaining polarity necessary for protein trafficking involving vesicle fusion and exocytosis. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

STX3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q08849, Q13277
Stx3
STX3 Fusion Protein Ag7805
150 μL
Primary
Human, Rat
Antibody
IgG1
Western Blot
1B9C7
Unconjugated
STX3
STX3, STX3A, syntaxin 3
Mouse
Protein G
RUO
6809, 81802
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.