Learn More
Sampling Systems™ Stainless Steel Tweezers
These stainless steel tweezers are essential equipment for laboratories.
Brand: Sampling Systems™ A394-100
Description
- Polished finish
- Two sizes available
- Made from hygienic 304 grade stainless steel
Specifications
Stainless Steel (304) | |
100 mm | |
Tweezers | |
General laboratory use |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Sampling Systems™ Stainless Steel Tweezers > Length: 100mm