Learn More
Invitrogen™ Avidin, FITC conjugate
Click to view available options
Unit Size:
5mg
Each
Packaging:
1 Each
Description
Our avidin, fluorescein conjugate has excitation and emission maxima ∼494/518. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
Specifications
Specifications
| Conjugate | FITC (Fluorescein) |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Form | Liquid |