Learn More
Bandelin Electronic™ Sonorex™ Super RK Series Heated Ultrasonic Baths
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Heated Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
Brand:  Bandelin Electronic™ 316
 View more versions of this product
Product Code. 10334251
  • € 1755.00 / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Description
  • SweepTec™ for permanent sound field oscillation
  • LCT-transducers - working frequency of 35 kHz
  • All units are CE-marked
  • Degree of protection IP 32
  • Timer for 1-15 minutes or continuous operation
  • With thermostatically adjustable heater, 30°-80°C
  • Excellent for technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Specifications
2 A
230 V
300 x 150 x 150 mm (LxWxD)
Stainless Steel
5.5 L
RK 255H