missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bandelin Electronic™ Sonorex™ Super RK Series Heated Ultrasonic Baths
Provides for intense yet gentle cleaning of technical parts. Bandelin Electronic™ Sonorex™ Super RK Series Heated Ultrasonic Baths feature tanks and drip-proof housings of stainless steel.
Marca: Bandelin Electronic™ 263
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
- SweepTec™ for permanent sound field oscillation
- LCT-transducers - working frequency of 35 kHz
- All units are CE-marked
- Degree of protection IP 32
- Timer for 1-15 minutes or continuous operation
- With thermostatically adjustable heater, 30°-80°C
- Excellent for technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
18.7 A | |
230 V | |
325 x 300 x 200 mm (LxWxD) |
Stainless Steel | |
187 L | |
RK 514BH |