Learn More
Amarell™ Standard Density Hydrometer without Thermometer, measuring span 0.060g/cm3
Calibrated for reference temperature 20°C. The standard density hydrometer without thermometer can be used for many general specific gravity measurements.
Brand:  Amarell™ H800024
 View more versions of this product
Product Code. 10261822
Description
Description
  • Reference temperature 20°C
  • Each hydrometer length is 160mm
Specifications
Specifications
1Each
±1g/cm3
160 mm
20°C
Density Hydrometer
0.001g
1.300 to 1.360 g/cm3
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Amarell™ Standard Density Hydrometer without Thermometer, measuring span 0.060g/cm3 > Range: 1.300 to 1.360g/cm3