Learn More
Lenz Laborglasintrumente Silicone Sealing Rings
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
€ 125.00 - € 213.00
Product Code | Brand | Bore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bore Size | Price | Quantity & Availability | |||||
11536894
|
Lenz Laborglasintrumente
01333216 |
16 mm |
€ 125.00
Pack of 10 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
10060341
|
Lenz Laborglasintrumente
01334532 |
32 mm |
€ 213.00
Pack of 10 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
11516894
|
Lenz Laborglasintrumente
01333212 |
12 mm |
€ 135.00
Pack of 10 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
11546894
|
Lenz Laborglasintrumente
01333218 |
18 mm |
€ 128.00
Pack of 10 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
11506894
|
Lenz Laborglasintrumente
01333210 |
10 mm |
€ 137.00
Pack of 10 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
11526894
|
Lenz Laborglasintrumente
01333214 |
14 mm |
€ 134.00
Pack of 10 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
Description
- For screw caps with bore
- Material: silicone rubber (VMQ)
- With bonded PTFE face
Specifications
Silicone |