Learn More
Lenz Laborglasintrumente Silicone Sealing Rings
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01333214
This item is not returnable.
View return policy
Description
- For screw caps with bore
- Material: silicone rubber (VMQ)
- With bonded PTFE face
Specifications
14 mm | |
For thread 32 GL | |
29 mm | |
Silicone |