Learn More
Ambion™ Silencer™ Custom Designed siRNA
Click to view available options
Purity:
HPLC
Standard
Quantity:
20 nmol
40 nmol
5 nmol
Unit Size:
20nmol
40nmol
5nmol
Each
Description
- Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
- Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
Specifications
Spezifikation
| Content And Storage | Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water. |
| Description | GeneAssist - Custom Designed Silencer™ siRNA |
| For Use With (Application) | Knocking down gene expression in order to study protein function in a wide range of cell types |
| Product Line | Silencer |
| Product Type | Custom Designed siRNA |
| Purity | Standard |
| Quantity | 5 nmol |
| Shipping Condition | Ambient |
| Temperature (Metric) | Upon receipt, store in a non-frost-free freezer at or below -20°C |
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur