Learn More
Endecotts™ Sieve Calibration Ball
Sieve balls, used to improve the sieving of cohesive material
€ 260.00 - € 428.00
Specifications
Product Type Microspheres
Material Soda-lime glass
For Use With (Equipment) Sieve
View More Specs
Products 2
Product Code Brand Particle Size Price Quantity & Availability  
Product Code Brand Particle Size Price Quantity & Availability  
11924315
View Documents
Endecotts™
ZSICSA-1.40
14 mesh
€ 428.00
Pack of 3
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
11934295
View Documents
Endecotts™
ZSICSA-.038
400 mesh
€ 260.00
Pack of 5
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
Description
Description
    • Spheres are traceable to the United Kingdom National Physical Laboratory
    • Calibrate sieves to an accuracy of 1μm
    • Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture
    • Standards include a calibration chart
Specifications
Specifications
Microspheres
Soda-lime glass
Sieve