Learn More
Endecotts™ Sieve Calibration Ball
Sieve balls, used to improve the sieving of cohesive material
Brand:  Endecotts™ ZSICSA-1.40
 View more versions of this product
Product Code. 11924315
  • € 428.00 / Pack of 3
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
    • Spheres are traceable to the United Kingdom National Physical Laboratory
    • Calibrate sieves to an accuracy of 1μm
    • Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture
    • Standards include a calibration chart
Specifications
Specifications
Microspheres
Soda-lime glass
Sieve
14 mesh
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers