Learn More
Lenz Laborglasintrumente Separating Funnels, Conical, Amber, without Scale, with PTFE Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
€ 172.00 - € 351.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11903451
|
Lenz Laborglasintrumente
04008173 |
2 L |
€ 254.00
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
11913451
|
Lenz Laborglasintrumente
04008170 |
1 L |
€ 351.00
Pack of 2 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
11963441
|
Lenz Laborglasintrumente
04008128 |
50 mL |
€ 172.00
Pack of 2 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
11973441
|
Lenz Laborglasintrumente
04008137 |
100 mL |
€ 186.00
Pack of 2 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
11993441
|
Lenz Laborglasintrumente
04008158 |
500 mL |
€ 269.00
Pack of 2 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
- With NS socket and PE stopper
- Without scale, with PTFE plug
- With screwthread retaining device
- Material: borosilicate glass 3.3
Specifications
Borosilicate Glass (3.3) |