Learn More
Lenz Laborglasintrumente Separating Funnels, Conical, Amber, without Scale, with PTFE Plug
Click to view available options
Capacity (Metric):
50 mL
100 mL
500 mL
1 L
2 L
Unit Size:
Each
Pack of 2
Description
- With NS socket and PE stopper
- Without scale, with PTFE plug
- With screwthread retaining device
- Material: borosilicate glass 3.3
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Material | Borosilicate Glass (3.3) |