Learn More
Cytiva Sentino™ Supor™ Membrane
Filter dispense packs for Sentino dispenser are ideal for high throughput microbiology testing labs.
Brand: Cytiva 68123
This item is not returnable.
View return policy
Description
- Ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes
- Gamma irradiated and ready to use
- For research use only
Specifications
PES | |
0.2 μm | |
Non-sterile | |
1000/Pk., Dispenser Pack of 200 |
Product Content Correction
Product Title
Cytiva Sentino™ Supor™ Membrane > 0.2 μm