Learn More
Brady™ Roll of Not Drinking Water Warning Labels
Brand: Brady™ 227410
This item is not returnable.
View return policy
Specifications
PIC 204 RL-DIA 025-B7541 | |
Laminated Polyester | |
Permanent Acrylic | |
0.099 mm | |
Round | |
Black on White |
Gloss | |
250/Pk. | |
25 mm | |
-40 °C-110 °C | |
Global |