Learn More
Emma Safety Footwear Rocky Safety Shoes
The go-anywhere safety shoe that molds itself to your foot. Uppers are crafted from soft, supple nubuck leather with anti-slip soles. Suited to all aspects of loading and unloading, climbing onto palette stacks, and getting in and out of truck cabs whatever the weather.
Brand:  Emma Safety Footwear MM72054642
 View more versions of this product
Product Code. 13216354
  • € 192.00 / 1 pair
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Description
  • S3-Safety shoe with sole plate for wet work
  • Nubuck / Cordura™ construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Gray
Hydro-Tec™ Sanitized Silver
42
Laces
Rocky
Cordura
Steel