Learn More
Epredia™ Decalcifying Solution
Remove calcium from histological specimens with Epredia™ Decalcifying Solution.
Brand:  Epredia™ 8340
 View more versions of this product
Product Code. 12697766
Description
Description
  • Offers rapid decalcification without destroying nuclear properties of the tissue
  • The EDTA and dilute hydrochloric acid in this solution require no heat, electricity, or other aids to decalcify bone tissue
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Decalcifying Solution
32 oz.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Decalcifying Solution > 32 oz.