Learn More
Labconco™ ReVo Canopy Connection Kit
ReVo Canopy Connection Kit is ideal for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
€ 1305.00 - € 1410.00
Specifications
Product Type | Canopy Connection Kit |
---|---|
Dimensions (L x W x H) Exterior | 465 x 680 x 265 mm |
Shipping Weight (Metric) | 5.0 kg |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17805602
|
Labconco™
R3889200 |
900/1200 mm MSC |
€ 1305.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17845602
|
Labconco™
R3889202 |
1500/1800 mm Cabinet |
€ 1410.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Canopy Connection Kit with top exhaust for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Specifications
Canopy Connection Kit | |
465 x 680 x 265 mm | |
5.0 kg |
Product Title
Labconco™ ReVo Canopy Connection Kit