Learn More
Labconco™ ReVo Canopy Connection Kit
Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
1500/1800 mm Cabinet
900/1200 mm MSC
Unit Size:
1 set
Description
- Canopy Connection Kit with top exhaust for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Specifications
Specifications
| Product Type | Canopy Connection Kit |
| Dimensions (L x W x H) Exterior | 465 x 680 x 265 mm |
| For Use With (Equipment) | 1500/1800 mm Cabinet |
| Shipping Weight (Metric) | 5.0 kg |