Learn More
BUCHI FlashPure ID Silica Flash Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
€ 409.00
Specifications
Material Silica
For Use With (Application) Useful for Conventional Applications; Polar Compounds
View More Specs
Products 1
Product Code Brand Bed Weight (Metric) Price Quantity & Availability  
Product Code Brand Bed Weight (Metric) Price Quantity & Availability  
16360276
View Documents
BUCHI
145146133
80 g
€ 409.00
Pack of 12
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications
Specifications
Silica
Useful for Conventional Applications; Polar Compounds
Documents
Product Certifications
Product Content Correction
Product Title
BUCHI FlashPure ID Silica Flash Chromatography Cartridges