Learn More
BUCHI FlashPure ID Silica Flash Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
Brand:  BUCHI 145146133
 View more versions of this product
Product Code. 16360276
  • € 409.00 / Pack of 12
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Includes: 20 Pieces
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications
Specifications
Silica
12 Pc.
Useful for Conventional Applications; Polar Compounds
Reveleris
80 g
200 psig
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BUCHI FlashPure ID Silica Flash Chromatography Cartridges > FlashPure ID; Stationary Phase: Silica; Size: 80 g; Quantity: 12 pcs