missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Reinforced Polypropylene Mandatory Signs
Brand: Brady™ 252136
This item is not returnable.
View return policy
Specifications
PIC 267-DIA 315-B7527 | |
1 Ea. | |
315 mm | |
0.369 mm | |
Use Breathing Protection | |
White on Blue |
Polypropylene | |
Reinforced Polypropylene | |
120°C | |
Round | |
Mandatory Sign |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction