missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Screwthread Tubes for Glass Blowers

Manufactured from borosilicate glass for highest resistance to thermal shock and chemical corrosion. Quickfit™ Screwthread Tubes for Glass Blowers combined with plastic fittings produce a tight connection to a straight tube or provide an interface with other materials.

Brand:  Quickfit SQ13UB

56.67 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10355450

  • € 61.15 / Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Unbadged (free from ceramic inscriptions) to avoid charring when badge is subject to direct heat
  • External threading fits standard GL thread sizes
  • For use by Glassblowers in the construction of laboratory glassware
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Designed for use with a range of plastic screwcaps, silicone rubber rings, screwthread connectors, and PTFE washers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

135 mm
the construction of laboratory glassware
11.1 mm
Borosilicate Glass
GL 13
Screwthread Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers