missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Procyanidin B2
MedChemExpress Procyanidin B2 is a natural flavonoid, with anti-cancer, antioxidant activities.
Brand: MedChemExpress HY-N0796

This item is not returnable.
View return policy
Specifications
Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month | |
Room temperature | |
29106-49-8 | |
98.07% | |
Proanthocyanidin B2 | |
10 mg |
578.52 g/mol | |
Cancer-programmed cell death | |
Natural Products | |
H2O : 66.67 mg/mL (115.24 mM∣Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (86.43 mM) | |
Reactive Oxygen Species |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MedChemExpress Procyanidin B2
Spot an opportunity for improvement?Share a Content Correction