Learn More
Electrothermal™ Energy Regulator
For use with Electrothermal™ EM Series Electromantles. Bibby Scientific™ Electrothermal Energy Regulators enable an even and accurate transfer of heat.Includes indicator lamps for power and heater operation.
Brand: Electrothermal 10085-14
This item is not returnable.
View return policy
Description
- For use with Electrothermal™ EM Series Electromantles.
- Enables an even and accurate transfer of heat.
- Includes indicator lamps for power and heater operation.
Specifications
Solid state Simmerstat assembly | |
90/250 V | |
Energy Regulator |
Electrothermal™ EM Series Electromantles | |
Indicator lamps for power and heater operation |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Electrothermal™ Energy Regulator > For Use With: Heating Mantle