Learn More
Greiner Bio-One Polystyrene Conical Bottom Analyzer Cups
Greiner Bio-One Cell 1.5mL Polystyrene Conical Bottom Analyzer Cups suitable for use with analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analyzers.
Brand: Greiner Bio-One 624101
This item is not returnable.
View return policy
Description
- Material: Polystyrene
- Conical bottom
- With support skirt
- For single use only
Specifications
Gemsaec Analyzer | |
Non-sterile | |
Polystyrene | |
0.5 mL |
Conical | |
Clear | |
Analyzer Cup |