missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyanetholesulfonic Acid Sodium Salt, Reagent Grade, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Specifications
Specifications
Color | White |
Quantity | 25 g |
Formula Weight | (250)n |
Grade | Reagent |
Physical Form | Solid |
Chemical Name or Material | Polyanetholesulfonic Acid Sodium Salt |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Polyanetholesulfonic Acid Sodium Salt, Reagent Grade, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction