missing translation for 'onlineSavingsMsg'
Learn More

Pexidartinib, MedChemExpress

Encompass

MedChemExpress Pexidartinib (PLX-3397) is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor, with IC50s of 20 and 10 nM, respectively. Pexidartinib (PLX-3397) exhibits 10- to 100-fold selectivity for c-Kit and CSF1R over other related kinases. Pexidartinib (PLX-3397) induces cell apoptosis and has anti-tumor activity.

Encompass_Preferred

€ 78.30 - € 451.50

Specifications

Chemical Name or Material Pexidartinib
CAS 1029044-16-3
Color White
Purity Grade Notes Research
Molecular Formula C20H15ClF3N5
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16473197
Encompass_Preferred
View Documents
Medchem Express
HY-16749
50 mg
€ 249.75
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16467166
Encompass_Preferred
View Documents
Medchem Express
HY-16749
100 mg
€ 306.45
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16439707
Encompass_Preferred
View Documents
Medchem Express
HY-16749
5 mg
€ 78.30
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16437178
Encompass_Preferred
View Documents
Medchem Express
HY-16749
10 mg
€ 108.00
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16411236
Encompass
View Documents
Medchem Express
HY-16749
200 mg
€ 451.50
200mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pexidartinib (PLX-3397) is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor, with IC50s of 20 and 10 nM, respectively. Pexidartinib (PLX-3397) exhibits 10- to 100-fold selectivity for c-Kit and CSF1R over other related kinases. Pexidartinib (PLX-3397) induces cell apoptosis and has anti-tumor activity.
Specifications

Specifications

Pexidartinib
White
C20H15ClF3N5
DMSO : 100 mg/mL (239.34 mM; Need ultrasonic)
417.81
98.0%
Powder
1029044-16-3
Research
PLX-3397
FC(C1=CC=C(CNC2=NC=C(CC3=CNC4=NC=C(Cl)C=C43)C=C2)C=N1)(F)F
417.81
Research
Cancer-Kinase/protease
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Pexidartinib, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.