Learn More
Pexidartinib, MedChemExpress
MedChemExpress Pexidartinib (PLX-3397) is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor, with IC50s of 20 and 10 nM, respectively. Pexidartinib (PLX-3397) exhibits 10- to 100-fold selectivity for c-Kit and CSF1R over other related kinases. Pexidartinib (PLX-3397) induces cell apoptosis and has anti-tumor activity.
Brand:  Medchem Express HY-16749
Encompass_Preferred
 View more versions of this product
Product Code. 16439707
  • € 78.30 / 5mg
Explore more special offers
Description
Description
Pexidartinib (PLX-3397) is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor, with IC50s of 20 and 10 nM, respectively. Pexidartinib (PLX-3397) exhibits 10- to 100-fold selectivity for c-Kit and CSF1R over other related kinases. Pexidartinib (PLX-3397) induces cell apoptosis and has anti-tumor activity.
Specifications
Specifications
Pexidartinib
White
C20H15ClF3N5
PLX-3397
FC(C1=CC=C(CNC2=NC=C(CC3=CNC4=NC=C(Cl)C=C43)C=C2)C=N1)(F)F
417.81
Research
Cancer-Kinase/protease
1029044-16-3
Research
5 mg
DMSO : 100 mg/mL (239.34 mM; Need ultrasonic)
417.81
98.0%
Powder
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Pexidartinib, MedChemExpress
Research purposes only