Learn More
Thermo Scientific™ Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Designed for durability and fitness for purpose, a range of Thermo Scientific™ Peristaltic Pump Tubing for the iCAP™ MX and iCAP™ Q/Qnova Series is available to suit your sample matrices and ICP-MS applications.
€ 330.00
Specifications
Type | Peristaltic Pump Tubing |
---|---|
For Use With (Equipment) | iCAP MX and iCAP Q/Qnova Series |
Description
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.
Select High Quality Consumables for Improved Performance
Choose from the following Peristaltic Pump Tubing:
- Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
- Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
- Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
- Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
- Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
- Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene
Specifications
Peristaltic Pump Tubing | |
iCAP MX and iCAP Q/Qnova Series |