Learn More
Thermo Scientific™ Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Designed for durability and fitness for purpose, a range of Thermo Scientific™ Peristaltic Pump Tubing for the iCAP™ MX and iCAP™ Q/Qnova Series is available to suit your sample matrices and ICP-MS applications.
Brand:  Thermo Scientific™ 1320120
 View more versions of this product
Product Code. 16533192
  • € 330.00 / Each
Explore more special offers
Description
Description
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.
Select High Quality Consumables for Improved Performance
Choose from the following Peristaltic Pump Tubing:
  • Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
  • Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
  • Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
  • Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
  • Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
  • Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene
Specifications
Specifications
Peristaltic Pump Tubing
Santoprene, Dia. 3.175, Black/White
iCAP MX and iCAP Q/Qnova Series
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System >