missing translation for 'onlineSavingsMsg'
Learn More

Peramivir trihydrate, MedChemExpress

MedChemExpress Peramivir trihydrate (RWJ-270201 trihydrate;BCX-1812 trihydrate) is a highly potent, selective and orally active influenza virus neuraminidase (NA) inhibitor, with IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes.

€ 214.50 - € 607.50

Specifications

Chemical Name or Material Peramivir trihydrate
Physical Form Powder
CAS 1041434-82-5
Color White
Purity Grade Notes Research
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16457268
Encompass
View Documents
Medchem Express
HY-17015
10 mg
€ 214.50
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16413287
Encompass_Preferred
View Documents
Medchem Express
HY-17015
50 mg
€ 607.50
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Peramivir trihydrate (RWJ-270201 trihydrate
Specifications

Specifications

Peramivir trihydrate
1041434-82-5
Research
COVID-19-anti-virus
H2O : 5 mg/mL (13.07 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
382.45
98.0%
Powder
White
C15H34N4O7
RWJ 270201 trihydrate BCX 1812 trihydrate
O[C@H]1[C@]([C@H](C(CC)CC)NC(C)=O)([H])[C@H](NC(N)=N)C[C@@H]1C(O)=O.[H]O.[H]O.[H]O
382.45
Research
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.