Learn More
Peramivir trihydrate, MedChemExpress
MedChemExpress Peramivir trihydrate (RWJ-270201 trihydrate;BCX-1812 trihydrate) is a highly potent, selective and orally active influenza virus neuraminidase (NA) inhibitor, with IC50 values ranging from 0.9 to 4.3 nM for nine NA subtypes.
Brand:  Medchem Express HY-17015
Encompass_Preferred
 View more versions of this product
Product Code. 16413287
  • € 607.50 / 50mg
Explore more special offers
Description
Description
Peramivir trihydrate (RWJ-270201 trihydrate
Specifications
Specifications
Peramivir trihydrate
1041434-82-5
50 mg
C15H34N4O7
RWJ 270201 trihydrate BCX 1812 trihydrate
O[C@H]1[C@]([C@H](C(CC)CC)NC(C)=O)([H])[C@H](NC(N)=N)C[C@@H]1C(O)=O.[H]O.[H]O.[H]O
382.45
Research
Powder
White
Research
COVID-19-anti-virus
H2O : 5 mg/mL (13.07 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
382.45
98.0%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Peramivir trihydrate, MedChemExpress
Research purposes only