missing translation for 'onlineSavingsMsg'
Learn More

PD 0325901, Tocris Bioscience™

Product Code. 18095742
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent inhibitor of MEK1/2

PD 0325901 is a potent MEK1 and MEK2 inhibitor. PD 0325901 inhibits MEK activity in mouse colon 26 cells (IC50 = 0.33 nM). PD 0325901 inhibits the growth of melanoma cell lines in vitro and in vivo; induces G1-phase cell cycle arrest and apoptosis in a mouse xenograft model. Also inhibits production of proangiogenic cytokines such as VEGF. Enhances generation of induced pluripotent stem cells (iPSCs). Orally active.

For more information about how PD 0325901 may be used, see our protocols: Culturing Transcriptomically Distinct Pluripotent mESCs, Highly Efficient Generation of CiPSCs from MEFs, Accelerated Induction of Cortical Neurons from hiPSCs .

Chemical Identifiers

CAS 391210-10-9
Molecular Formula C16H14F3IN2O4
Molecular Weight (g/mol) 482.198
InChI Key SUDAHWBOROXANE-SECBINFHSA-N
Synonym r-n-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, r-n-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenylamino benzamide, unii-86k0j5ak6m, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, n-2r-2,3-dihydroxypropyl oxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, n-r-2,3-dihydroxy-propoxy-3,4-difluoro-2-2-fluoro-4-iodo-phenylamino-benzamide, n-r-2,3-dihydroxypropyl oxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino benzamide, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodoanilino benzamide, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodophenyl amino-benzamide, n-2r-2,3-dihydroxypropoxy-3,4-difluoro-2-2-fluoro-4-iodo-anilino benzamide
PubChem CID 9826528
IUPAC Name N-[(2R)-2,3-dihydroxypropoxy]-3,4-difluoro-2-(2-fluoro-4-iodoanilino)benzamide
SMILES C1=CC(=C(C=C1I)F)NC2=C(C=CC(=C2F)F)C(=O)NOCC(CO)O

Specifications

Quantity 10 mg
Formula Weight 482.19
Percent Purity >99%
Chemical Name or Material PD 0325901
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'