Learn More
Thermo Scientific™ Oxoid™ Lincomycin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Brand:  Thermo Scientific™ CT0027B
 View more versions of this product
Product Code. 11974022
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Lincomycin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lincomycin
Oxoid Lincomycin Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
2μg
MY
Antimicrobial Susceptibility
50 Discs/Cartridge
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6103897 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog NumberCT0027B
Lot Number3552261 Certificate TypeCertificate of Analysis Date28/09/2022 Catalog NumberCT0027B
Lot Number3294953 Certificate TypeCertificate of Analysis Date28/05/2021 Catalog NumberCT0027B
Lot Number2975671 Certificate TypeCertificate of Analysis Date27/05/2020 Catalog NumberCT0027B
Lot Number2847578 Certificate TypeCertificate of Analysis Date04/09/2019 Catalog NumberCT0027B
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Lincomycin Antimicrobial Susceptibility discs >