Learn More
Thermo Scientific™ Oxoid™ Amoxycillin/Clavulanic Acid Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Brand:  Thermo Scientific™ CT0538B
 View more versions of this product
Product Code. 11913812
  • € 87.35 / Pack of 250
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Amoxycillin/Clavulanic Acid Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amoxycillin/Clavulanic Acid
3μg
AMC
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with BSAC, EUCAST guidelines
Oxoid Amoxycillin/Clavulanic Acid Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6230137 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog NumberCT0538B
Lot Number6191421 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog NumberCT0538B
Lot Number6191420 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog NumberCT0538B
Lot Number6172431 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog NumberCT0538B
Lot Number6172432 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog NumberCT0538B
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Amoxycillin/Clavulanic Acid Antimicrobial Susceptibility discs >