Learn More
Thermo Scientific™ Orion™ ISE Calibration Standards
Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.
Brand:  Thermo Scientific™ 951207
 View more versions of this product
Product Code. 12861521
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.
All standards are 1 pint (475mL) bottles, unless otherwise noted.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ammonia Standard
475 mL
C of A Acceptance Range: 100 ppm +/- 1 ppm as Nitrogen
Ammonia Ion Selective Electrodes Including 9512BNWP, 9512HPBNWP
100 ppm as N
Ammonia Ion Selective Electrode
Calibration Standard
1 Bottle
Yes
Yes
Liquid
Colorless
Calibration Standard
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCT1 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number951207
Lot NumberCZ1 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number951207
Lot NumberBW1 Certificate TypeCertificate of Analysis Date26/05/2023 Catalog Number951207
Lot NumberAS1 Certificate TypeCertificate of Analysis Date09/09/2022 Catalog Number951207
Lot NumberAS1 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog Number951207
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ ISE Calibration Standards > Ammonia Standard, 100ppm as Nitrogen (N), 475mL
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.