Learn More
Brady™ Nylon Wire & Cable Ties
Cable ties constructed of nylon.
Brand: Brady™ 196311
This item is not returnable.
View return policy
Description
- Nylon
- 200 mm long
Specifications
Nylon | |
NYTIE-3.5X200-NT | |
White | |
200 x 3.5 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Brady™ Nylon Wire & Cable Ties > Color: White