Learn More
Brady™ Nylon Wire & Cable Ties
Cable ties constructed of nylon.
Brand: Brady™ 196351
This item is not returnable.
View return policy
Description
- Nylon
- 200 mm long
Specifications
Nylon | |
NYTIE-3.6x200-RD | |
Red | |
200 x 3.6 mm |
Product Content Correction
Product Title
Brady™ Nylon Wire & Cable Ties > Color: Red