Learn More
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Click to view available options
No. of Wells:
96
384
No. per Pack:
1 Microplates
10 Microplates
25 Microplates
No. per Case:
100 Microplates
30 Microplates
Description
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.
- Clear Polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Customized bar coding is available
- Flexible working volumes; 10-100μL
Specifications
Specifications
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Format | Standard |
Lid | With Lid |
Culture Area | 0.084 cm2/well |
Description | Non-Treated; w/ lid; Sterile; 10/30 |
Array | 16 x 24 |
Packaging | 10 Pk., 30 Cases |
Product Line | Nunc |
Color | Clear |
Show More |
Documents
Product Information
Manuals
Technical Data Sheet: Nunc 384-Well, PS, Flat Bottom Plate
Version: August 2015
File size: 104.8 kB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
184770 | Certificate of Irradiation | 20/01/2025 | 242757 |
184770 | Certificate of Compliance | 04/01/2025 | 242757 |
1326977 | Certificate of Compliance | 11/12/2024 | 242757 |
1417524 | Certificate of Processing | 06/12/2024 | 242757 |
1406155 | Certificate of Processing | 04/12/2024 | 242757 |
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate >