missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Click to view available options
No. of Wells:
96
384
No. per Pack:
1 Microplates
10 Microplates
25 Microplates
No. per Case:
100 Microplates
30 Microplates
Description
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.
- Clear Polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Customized bar coding is available
- Flexible working volumes; 10-100μL

Specifications
Specifications
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | With Lid |
Culture Area | 0.084 cm2/well |
Description | Non-Treated; w/ lid; Sterile; 1/30 |
Array | 16 x 24 |
Packaging | 1/Pk., 30/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Show More |
Documents
Product Information
Manuals
Technical Data Sheet: Nunc 384-Well, PS, Flat Bottom Plate
Version: August 2015
File size: 104.8 kB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1290545 | Certificate of Compliance | 11/12/2024 | 242765 |
1375940 | Certificate of Processing | 04/12/2024 | 242765 |
1303183 | Certificate of Processing | 04/12/2024 | 242765 |
1337196 | Certificate of Processing | 04/12/2024 | 242765 |
1361112 | Certificate of Processing | 04/12/2024 | 242765 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate >
Spot an opportunity for improvement?Share a Content Correction