Learn More
NQ301, MedChemExpress
MedChemExpress NQ301 is an antithrombotic agent; inhibits collagen-challenged rabbit platelet aggregation with an IC50 of 10 mg/mL.
Brand: Medchem Express HY-101054
This item is not returnable.
View return policy
Description
NQ301 is an antithrombotic agentSpecifications
NQ301 | |
Red | |
C18H12ClNO3 | |
DMSO : ≥ 29 mg/mL (89.03 mM) | |
325.75 | |
98.12% | |
Solid |
130089-98-4 | |
Research | |
100 mg | |
O=C1C(NC2=CC=C(C(C)=O)C=C2)=C(Cl)C(C3=C1C=CC=C3)=O | |
325.75 | |
Research | |
COVID-19-immunoregulation |
Research purposes only