Learn More
NQ301, MedChemExpress
Product Code. 16453248
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16453248
Brand: Medchem Express HY101054
MedChemExpress NQ301 is an antithrombotic agent; inhibits collagen-challenged rabbit platelet aggregation with an IC50 of 10 mg/mL.
NQ301 is an antithrombotic agent
Specifications
Chemical Name or Material NQ301
CAS 130089-98-4
Color Red
Purity Grade Notes Research
Molecular Formula C18H12ClNO3
Quantity 10 mg
Solubility Information DMSO : ≥ 29 mg/mL (89.03 mM)
SMILES O=C1C(NC2=CC=C(C(C)=O)C=C2)=C(Cl)C(C3=C1C=CC=C3)=O
Molecular Weight (g/mol) 325.75
Formula Weight 325.75
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only