Learn More
NQ301, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
NQ301 is an antithrombotic agent
Specifications
Specifications
| Chemical Name or Material | NQ301 |
| CAS | 130089-98-4 |
| Color | Red |
| Purity Grade Notes | Research |
| Molecular Formula | C18H12ClNO3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 29 mg/mL (89.03 mM) |
| SMILES | O=C1C(NC2=CC=C(C(C)=O)C=C2)=C(Cl)C(C3=C1C=CC=C3)=O |
| Molecular Weight (g/mol) | 325.75 |
| Formula Weight | 325.75 |
| Show More |
Research purposes only