Learn More
Netilmicin sulfate, MedChemExpress
MedChemExpress Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.
€ 112.50 - € 600.75
Specifications
Chemical Name or Material | Netilmicin sulfate |
---|---|
CAS | 56391-57-2 |
Color | Yellow |
Purity Grade Notes | Research |
Molecular Formula | C21H41N5O7.5/2H2SO4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16484177
|
Medchem Express
HY-A0086 |
50 mg |
€ 411.75
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460728
|
Medchem Express
HY-A0086 |
5 mg |
€ 112.50
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16458026
|
Medchem Express
HY-A0086 |
100 mg |
€ 600.75
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16408298
|
Medchem Express
HY-A0086 |
10 mg |
€ 157.95
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.Specifications
Netilmicin sulfate | |
Yellow | |
C21H41N5O7.5/2H2SO4 | |
H2O : 100 mg/mL (138.74 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.39 mM; ultrasonic and warming and heat to 80°C) | |
720.78 | |
98.0% | |
Solid |
56391-57-2 | |
Research | |
SCH-20569 sulfate | |
NCC1=CC[C@@H](N)[C@](O[C@@H]2[C@@H](N)C[C@@H](NCC)[C@H](O[C@@]3([H])OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)([H])O1.[2.5H2SO4] | |
720.78 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H302∣H315∣H319∣H332∣H335∣H350∣H360
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.